missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Hardened Steel Grinding Sets
2255.00 € - 3870.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10796671
|
RETSCH
01.462.0171 |
100 mL |
2850.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
10039332
|
RETSCH
01.462.0263 |
250 mL |
3870.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
10212741
|
RETSCH
01.462.0170 |
50 mL |
2255.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Specifications
Grinding Set | |
Hardened Steel |