Learn More
Hanna Instruments™ Hardness Reagent Water Testing Kit
Evaluate the level of water hardness with this complete chemical test kit. Hanna Instruments™ Hardness Reagent Water Testing Kit measures total hardness as CaCO3 by titration with EDTA.
Brand:  Hanna Instruments™ HI3812
Product Code. 10327393
  • 181.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pre-made reagents for ease of use
  • All reagents marked with expiration date and lot number for traceability
  • Manual titration performed with calmagite indicator
Specifications
Specifications
100
0 to 300mg/L
Hardness Test Kit
1 Each
Titration
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products