Learn More
Sartorius Hardware Package for Built-in Ionizer
Activation Ionizer accessory for Cubis™ II for effectively neutralizing charges from samples, containers, or powders.
Brand:  Sartorius QAPP1001
Product Code. 17987320
  • 564.00 € / Each
Explore more special offers
Description
Description
  • Built-in Ionizer Upgrade for Sartorius Cubis II Ultra Hi-Resolution High-Capacity Micro Balances
  • Effectively neutralizes electrostatic charges from samples, container, or powder
  • Helps ensure accurate results and stable readouts
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cubis™ II High-Capacity Micro Balance
Software License