Learn More
Brady™ Harsh Environment Multipurpose Continuous Clear Polyester Labels for M6/M7 Printers
Get aggressive adhesive labels for difficult surfaces, and the harshest environments. These clear, polyester labels are ideal for labs, asset tracking and faceplate labeling.
Marque:  Brady™ 174332
 Afficher plus de versions de ce produit
Code produit. 17839014
  • 58.50 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Découvrez plus d'offres spéciales
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
  • High-tack permanent adhesive for stainless steel, powder coated metals, painted enamel, and plastics in all-weather applications
  • Resists exposure to UV light, extreme temperatures, fuel and oil, degreasers, cleaners, and detergents
  • Black print on clear polyester
  • UL Recognized to UL969, CSA to C222 No 015-95, RoHS compliant to 2011/65/EU (Pass 2015/863)
Spécification
Spécification
Autoclavable
Permanent Acrylic
Clear
Continuous
Single-side Printable
Labels
Asset Tracking, Barcode Labeling, Equipment Labeling, General Identification, Laboratory Labeling
No
Rectangle
-70°C to +100°C
0.076 mm
50 ft.
0.375 in.
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognized
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
50 ft.
-94°F to +212°F
0.003 in.
50 ft. x 0.375 in.
15.24 m
9.53 mm
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats