Learn More
Brady™ Harsh Environment Multipurpose Continuous Clear Polyester Labels for M6/M7 Printers
Get aggressive adhesive labels for difficult surfaces, and the harshest environments. These clear, polyester labels are ideal for labs, asset tracking and faceplate labeling.
Brand:  Brady™ 173297
 View more versions of this product
Product Code. 17839054
  • 95.35 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High-tack permanent adhesive for stainless steel, powder coated metals, painted enamel, and plastics in all-weather applications
  • Resists exposure to UV light, extreme temperatures, fuel and oil, degreasers, cleaners, and detergents
  • Black print on clear polyester
  • UL Recognized to UL969, CSA to C222 No 015-95, RoHS compliant to 2011/65/EU (Pass 2015/863)
Specifications
Specifications
Autoclavable
Permanent Acrylic
Clear
Continuous
Single-side Printable
Labels
Asset Tracking, Barcode Labeling, Equipment Labeling, General Identification, Laboratory Labeling
No
Rectangle
-70°C to +100°C
0.076 mm
50 ft.
1 in.
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognized
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
50 ft.
-94°F to +212°F
0.003 in.
50 ft. x 1 in.
15.24 m
25.4 mm