missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ HATU Coupling Reagent

Product Code. 15920663 Shop All Applied Biosystems Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15920663

Brand: Applied Biosystems™ GEN076525

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This is a coupling reagent for use in peptide synthesis.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
For Use With (Application) Peptide Synthesis
Product Type Peptide Synthesis Resin
Quantity 100 g
Shipping Condition Wet Ice
Form Resin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.