missing translation for 'onlineSavingsMsg'
Learn More

Hazard Label PIC W009 ISO Safety Symbol 'Warning, Biological Hazard' 22mm x 25mm

Laminated polyester, suitable for indoor or outdoor use

Brand:  Brady 827646

74.93 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 15358437

  • 92.50 € / 1 roll
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Brady labels and label printers, benchtop or handheld, offer a full range of capabilities and options to provide high quality, cost-effective printing on a complete line of materials.
For Hazardous Substances Identification (HSID). Safety at the work place has now become more important than ever. Hazardous working substances present a constant source of danger. Reduce the risk with these symbols from Brady.
Specifications

Specifications

ISO 7010:2011
Permanent Acrylic
Gloss
Indoor & Outdoor
250/Pk.
-40°C to +110°C
25 mm
PIC W009-Tri 025-PE-ROLL/1
Black on Yellow
Laminated Polyester
Global
Triangle
0.099 mm
22.00 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.