Learn More
3M™ Hazard Warning Tape
Used for marking hazardous areas, protruding equipment, or preventing accidents in factories, warehouses and hospitals.
Brand:  3M™ 7100015263
 View more versions of this product
Product Code. 17267790
  • 265.00 € / 24 rolls
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Use for taping in all non-critical applications
  • Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas
TRUSTED_SUSTAINABILITY
Specifications
Specifications
766 i
33 m
Black with Yellow Stripes
50 mm x 33 m
50 mm