Learn More
3M™ Hazardous Spill Response Kit
Contains chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. 3M™ Hazardous Spill Response Kits can be used to contain and clean up the hazardous liquid without delay.
121.00 € - 269.00 €
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Includes | Price | Quantity & Availability | |||
12327979
|
3M™
7100090073 |
26 L | 500 x 630 mm | 10 sheets, 5 pillows, 5 mini-booms and 2 disposable bags with ties. | N/A | ||||
12372459
|
3M™
HSRK-5 |
5 L | 400 x 70 mm | 10 sheets, 1 pillow, 1 disposable bag with tie | N/A | ||||
Description
Kits contain chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. In the event of a spill, the kit can be used to contain and clean up the hazardous liquid without delay.- Constructed from polypropylene fibers
- Available in a wide range of formats
- Lightweight and dust-free
- High absorption capacity minimizes waste for disposal.
Specifications
Polypropylene Fiber | |
Spill Kit | |
1 Ea. |