missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazardous Substance Labels, 76 mm Core

Product Code. p-7225705
Click to view available options
Includes:
1 Diamond Layout
2 Diamond Layout
3A Diamond Layout
3B Diamond Layout
4A Diamond Layout
4B Diamond Layout
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 15941021

Brand: Brady™ 622283

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pre-printed, benchtop printer labels for CLP-GHS hazardous substances.

  • Supplied with pre-printed red diamonds
  • Ready for you to print the required warning, compliant to the latest regulations

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Gloss
Format Die-cut
Material Polypropylene (B-7594)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
For Use With (Equipment) i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Includes 1 Diamond Layout
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model THTCLP-M-L1-7594-1
No. per Pack 1000 Label(s) / Roll
Quantity 1000 Labels
Shape Rectangle
Size 75.00 mm (W) x 110.00 mm (H)
Temperature Range (English) -4°F to 212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Vertical Repeat (Metric) 115.4 mm
Dimensions (LxW) 75.00 mm (W) x 110.00 mm (H)
Length (English) 4.33 in.
Width (English) 2.95 in.
Width (Metric) 75 mm
Width (English) Printable Area 75.00 mm
Height (English) 4.33 in.
Height (Metric) 110 mm
Alternate Part Number THTCLP-M-L1-7594-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.