missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Extendable Rack

The Hazardous Substances Rack provides storage of aggressive chemicals.

Brand:  asecos™ 27856

 View more versions of this product

Product Code. 17876526

  • 791.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars.
  • Storage levels in 25 mm increments, height-adjustable.
  • Load capacity per storage shelf: 200 kg (evenly distributed).
  • Flat-pack delivery with assembly instructions
  • Polyethylene sumps with high resistance against aggressive chemicals (such as acids and alkalis).
Specifications

Specifications

10.6 gal.
Steel
51.65 in.
51.65 in.
78.74 in.
25.08 in.
125.66 lb.
40 L
Drum Rack
1312.00 mm
1312.00 mm
2000.00 mm
637.00 mm
57 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.