missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Hazardous Substances Rack
The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.
Product Code | Brand | Capacity (Metric) | Weight (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Weight (Metric) | Price | Quantity & Availability | ||||
17816526
|
asecos™
27837 |
20 L | 55 kg |
N/A
|
N/A | ||||
17278450
|
asecos™
27841 |
25 L | 73 kg |
N/A
|
N/A | ||||
17886476
|
asecos™
27839 |
30 L | 67 kg |
N/A
|
N/A | ||||
17886416
|
asecos™
27845 |
32 L | 66 kg |
N/A
|
N/A | ||||
17844625
|
asecos™
27843 |
40 L | 76 kg |
N/A
|
N/A | ||||
17896496
|
asecos™
27849 |
41 L | 95 kg |
N/A
|
N/A | ||||
17826576
|
asecos™
27847 |
45 L | 82 kg |
N/A
|
N/A | ||||
17864625
|
asecos™
27851 |
57 L | 94 kg |
N/A
|
N/A | ||||
17816586
|
asecos™
27869 |
205 L | 102 kg |
N/A
|
N/A | ||||
17896406
|
asecos™
27865 |
205 L | 110 kg |
N/A
|
N/A | ||||
17836396
|
asecos™
27871 |
225 L | 199 kg |
N/A
|
N/A | ||||
17886186
|
asecos™
27867 |
225 L | 217 kg |
N/A
|
N/A | ||||
Description
- Galvanised rack frames
- Sturdy construction due to reinforcement cross bars
- Storage levels in 25 mm increments, height-adjustable
- Load capacity per storage shelf: 200 kg (evenly distributed)
- Rack system can be extended individually
- Flat-pack delivery with assembly instructions
- Rack with galvanised shelf as storage levels
- Collecting sump
- 100 mm ground clearance for transport with pallet truck
Specifications
Steel | |
Drum Rack |