missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe™ Sharpsbin and Hazardous waste containers with yellow single lid
GREENER_CHOICE

Product Code. p-200022894
Click to view available options
Capacity (Metric):
6 L
12 L
50 L
Unit Size:
Each
Pack of 20
Pack of 30
This item is not returnable. View return policy
Follow instructions for safe and correct use

Product Code. 18742825

Brand: Woodsafe™ 1060212

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
Follow instructions for safe and correct use

Woodsafe is a cost-effective solution for reducing the use of fossil based plastic by 80% and CO2 emissions by 66%, documented in in a full LCA. Produced with residue from the forest industry using green energy. Available in sizes from 0.5 L to 50 L. Single and double lid versions.

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the Swedish forest industry
  • Lowers the use of Fossil-based plastic by 80%!
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be securely permanently closed for transportation to the final disposal
  • My Green Labs ACT Label
  • Follows same process as yellow bins and can be used to transport the same substances
  • Natural color and smell from wood - No migration of substances
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic

Specifications

Capacity (Metric) 6 L
Certifications/Compliance EN ISO 23907-1:2019. UN3291, ADR Approved
Closure Type Permanent closure
Closure Color Yellow
Capacity (English) 1.58 gal.
Color Brown, Yellow
Dimensions (L x W x H) 31.5 x 18.6 x 14.8 cm (12.4 x 7.32 x 5.83 in.)
Disposable Single-use
Quantity 30 Pk.
Stackable Yes
Size 31.5 x 18.6 x 14.8 cm
Sterility Non-sterile
Autoclavable Autoclavable
Includes Bin, Lid, and Label
For Use With (Application) Safe disposal of sharps and potentially infectious waste or medical waste
Packaging Cardboard
Material Biocomposite
Product Type Biohazard Waste Container
Show More Show Less

Lower climate impact and ACT label

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.