Learn More
New Pig™ HazMat Spill Kits
Brand:  New Pig™ RFL236
Encompass_Preferred
 View more versions of this product
Product Code. 12913625
  • 147.89 € / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Feature PIG absorbents proven for rapid response
  • Socks stop spreading spills; PIG HazMat Pads absorb quickly
  • Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bags help make cleanup easier
  • Bright yellow bag and large, color-coded label make the kit easy to locate
  • Strap for easy carrying and hanging
  • Compact and easy to store
Specifications
Specifications
79.49 L
Oils, Coolants, Solvents, Water