missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS (10X), no calcium, no magnesium, phenol red
GREENER_CHOICE

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Brand:  Gibco™ 14180046

Product Code. 11560476

  • 77.50 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This HBSS is modified as follows:

  • With: Glucose, Phenol Red
  • Without: Calcium, Magnesium, Sodium Bicarbonate

The complete formulation is available.

Concentrated forms of Gibco™ HBSS require pH adjustment and filtration at the time of preparation (see protocol for details).

TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2556353 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14180046
Lot Number2568328 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14180046
Lot Number2475213 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14180046
Lot Number2672049 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14180046
Lot Number2786698 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number14180046
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ HBSS (10X), no calcium, no magnesium, phenol red > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.