Learn More
Gibco™ HBSS, no calcium, no magnesium, no phenol red
GREENER_CHOICE
HBSS, no calcium, no magnesium, no phenol red
Brand:  Gibco™ 14175053
 View more versions of this product
Product Code. 11530476
  • 29.19 € / 500mL
Edge
Explore more special offers
Description
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485 standards, FDA
Hanks' Balanced Salt Solution, 500mL, Liquid, 1 X Concentration, 6.7 to 7.8 pH, With Glucose, Sodium Bicarbonate, 36 Months Shelf Life, For Mammalian Cell Culture
Liquid
Glucose, Sodium Bicarbonate
Mammalian Cell Culture
500 mL
15 to 30°C
1 X
Hanks' Balanced Salt Solution
No Calcium, No Magnesium, No Phenol Red
6.7 to 7.8
For in-vitro diagnostic use