Learn More
hCG alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25014-1-AP-20UL
 View more versions of this product
Product Code. 16863535
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Luteinizing hormone (LH, also known as lutropin) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common alpha-subunit but also a specific beta-subunit, which characterizes each of these hormones.
Specifications
Specifications
hCG alpha
Polyclonal
Unconjugated
CGA
CG ALPHA, CGA, Choriogonadotropin alpha chain, Follitropin alpha chain, FSH alpha, FSHA, GPHa, GPHA1, HCG, hCG alpha, LHA, LSH alpha, Lutropin alpha chain, Thyrotropin alpha chain, TSH alpha, TSHA
Rabbit
Antigen Affinity Chromatography
RUO
1081
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01215
CGA
hCG alpha Fusion Protein Ag17313
20 μL