missing translation for 'onlineSavingsMsg'
Learn More

HCl Dilution Matrix, EP Grade, Reagecon™

Product Code. 12814733 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured according to European Pharmacopoeia Chapter 2. HCl Dilution Matrix, EP Grade, Reagecon™ is a volumetric solution of 10g/L hydrochloric acid. It is used as a solvent or dilution matrix for a variety of standards, particularly EP Primary, Standard and Reference color solutions.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 2 guidelines
  • Produced in accordance with ASTM, APHA, ACS, EP and USP methods
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use solution eliminates the time and labor of preparing precise standard solutions

Specifications

Concentration 10g/L HCl
CAS 7647-01-0, 7732-18-5
Quantity 1 L
Color Colorless
Certificate of Analysis Certificate of Analysis
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
Synonym Muriatic acid, Hydrogen chloride
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Grade Eu. Ph. Chapter 2
Odor Characteristic
Physical Form Liquid
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.