Learn More
Lonza™ HCM™ SingleQuots™ Kit
Supplements and growth factors kit for use with primary hepatocyte basal medium.
Brand:  Lonza™ CC-4182
Product Code. 11625510
  • 390.91 € / Each
Explore more special offers
Description
Description
  • Separately sold SingleQuot™ kit for supplementing basal medium (CC-3199).
  • Combined with basal medium, these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species.
  • The final medium is serum-free
  • Includes the following:
    • Transferrin
    • Ascorbic Acid Solution
    • HEGF
    • Recombinant Human Insulin (0.5%)
    • Hydrocortisone
    • BSA, Fatty Acid Free
    • GA-1000
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transferrin, Ascorbic Acid, HEGF, Recombinant Human Insulin (0.5%), Hydrocortisone, Fatty Acid-Free BSA, GA-1000
Hepatocyte Cells
Supplements and growth factors