Learn More
HDAC6 Mouse anti-Human, Unconjugated, Clone: 1C7C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67250-1-IG-20UL
This item is not returnable.
View return policy
Description
The HDAC6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC6. This antibody recognizes human antigen. The HDAC6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
HDAC6 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry | |
1C7C3 | |
Unconjugated | |
20 μL | |
10013 | |
IgG1 |