missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine Safety Waste Caps
Safety Waste Caps safely connect the pipework to the waste container. The integrated exhaust filter blocks solvent vapours – for the protection of health and the environment.
Brand: SymLine 306498
94.74 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Waste Disposal System | |
4 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm, 1 x tube connector for tubes with (ID) 5.0 - 11.5 mm | |
GL45 |
SymLine System | |
PTFE | |
Safety Waste Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction