missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Carboy with Stopcock

For Use With: storage, filling, transporting

Brand:  Kautex™ 2000084139

Product Code. 15931451

  • 353.00 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Natural color
  • Blue printed graduation markings
  • Screwed-on closure
  • Comes with an yellow dust cap (dust cap does not seal the carboy)
  • Stopcock connections are available as accessories
  • Applications include storage, transfer and transport
TRUSTED_SUSTAINABILITY
Specifications

Specifications

60 L
346 mm
Closure
2/Cs.
Carboy
50 mm
Storage, Filling, Transporting
HDPE
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers