Learn More
Sampling Systems™ SteriWare™ HDPE Disposable Powder Sampler
BIOPHARMA
Can be used straight from the bag. The SteriWare™ Powder Sampler is ideal for taking large volume samples from powders or granules.
Brand:  Sampling Systems™ 8240H-1000S
Product Code. 15200749
  • 377.00 € / Pack of 20
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Key Features:
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
Operation:
  • Push the sampler into the product. Ensuring he sampler is in the closed position.
  • Rotate handle to open up the slot.
  • After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler.
  • Remove sampler from the product.
  • Deposit the sample into a suitable container.
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable sampler, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Specifications
Specifications
Powder Sampler
100 mL
Sterile
For taking a large volume sample from a single known point
HDPE
Single-use
Conforms to FDA CFR 177.1520, EU Regulations 10/2011, EU Regulations 10/2011
987 mm