Learn More
Kartell™ HDPE Industrial Tank
Kartell™ HDPE Industrial Tank is useful in everyday laboratory practice and features a comfortable handle with a secure seal-cap following DIN recommendations.
Brand:  Kartell™ 0178100
 View more versions of this product
Product Code. 15706978
  • 366.00 € / Pack of 40
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features a standardized industrial packaging design
  • Useful in everyday laboratory practice
  • Made of High Density PE food grade
  • Includes a comfortable handle
  • Features a secure seal-cap following DIN recommendations
  • It's conform to UN regulation on transportation of dangerous and toxic liquids and is suitable for road (ADR-RID), sea (IMO-IMDG) and air (IATA-ICAO) transport
Specifications
Specifications
GL51
HDPE
30
Seal-cap
5 L