Learn More
Ratiolab™ HDPE Pasteur Pipet
Use these pipets for simple and safe liquid handling. Ratiolab™ HDPE Pasteur Pipets feature a low affinity surface which reduces the loss of cells and valuable proteins through binding.
Brand:  Ratiolab™ 2600206
 View more versions of this product
Product Code. 12623165
  • 35.15 € / Pack of 400
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • No risk of cross-contamination or carry-over
  • No danger of glass breakage with the resulting danger of infectious material escaping
  • Transparent polyethylene construction
  • Inert to biological fluids and most acids
  • Soft, elastic bulb
  • Burr-free orifice for neat, clean and precise dispensing
  • Clearly visible embossed graduations
  • BSE-free
  • Can be used for food
Specifications
Specifications
HDPE
Food analysis
6 mL
Pasteur Pipette
Non-sterile
400 Pack
148 mm