missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ HDPE, Rectangular Carboy with Spigot
GREENER_CHOICE

Product Code. 13438039 Shop All Fisherbrand Products
9 L
Click to view available options
Capacity (Metric):
9 L
20 L

For Use With: Ideal for storing and dispensing lab water and reagents; Color: White

Carboys Ideal for storing and dispensing lab water and reagents.

Ideal for storing and dispensing lab water and reagents

 

  • Durable construction for long service life
  • Molded graduations for 1 gallon and 51 increments
  • Choice of materials for general use or autoclavability
  • Space-saving rectangular design
  • Wide mouth closure allows for easy filling and cleaning access
  • Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
  • Spigot spout without the adapter fits 5/8-in. ID tubing
  • Materials meet FDA requirements for food use and USP VI criteria (non-toxic)

 

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.4 gal.
Capacity (Metric) 9 L
Certifications/Compliance FDA, USP VI
Color White
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Material HDPE
Mouth Style Wide
Product Type Carboy with Spigot
Shape Rectangular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ HDPE, Rectangular Carboy with Spigot >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.