missing translation for 'onlineSavingsMsg'
Learn More

Brand™ HDPE Soaking Jar

Manufactured from High-Density Polyethylene. BRAND™ HDPE Soaking Jars are used with pipet rinsing systems and draining racks.

Brand:  Brand™ 29210

 View more versions of this product

Product Code. 10707791

  • 140.00 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from High-Density Polyethylene.
  • Used with Pipet rinsing systems and draining racks.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

For 460mm L pipets
HDPE
240 mm Base
10 L
510 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers