missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HDPE Wide Neck Square Reagent Bottles

Product Code. 15475574
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2500 mL
4000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

The Wide-neck reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.

  • HDPE, transparent
  • PP tamper-evident screw cap, blue
  • UN approval for bulk goods

Specifications

Capacity (Metric) 50 mL
Color Transparent
Diameter (Metric) Outer Neck 28 mm
Dimensions (L x W x H) 36 x 36 x 89 mm
Diameter (Metric) Inner Neck 21 mm
For Use With (Application) Foodstuffs
Material HDPE
Neck ID (Metric) 21 mm
Weight (Metric) 0.018 kg
Width (Metric) 36 mm
Closure Color Blue
Closure Included Yes
Height (Metric) 89 mm
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'