Learn More
3M™ Attest™ Biological Indicators
Monitors the effectiveness of the steam sterilization process and provides results within 48 hours of incubation. 3M™ Attest™ Biological Indicators provide the best assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores.
Brand:  3M™ 7000002535
Product Code. 11973604
  • 446.00 € / Pack of 100
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easy to use and interpret with visual color readout
  • Self-contained biological indicators significantly reduce possibility of contamination
  • Minimizes false positives
Specifications
Specifications
Autoclavable
Pink to Brown
100 Pk.
EN886-1-2 et ISO 11138-1-3
Biological Indicator