missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Heart Tissue Slides (Thrombus)- Paraffin

Product Code. 18171292 Shop All Bio Techne Products
Click to view available options
Species:
Human
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 18171292

Brand: Novus Biologicals™ NBP230267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Type Heart Tissue Slides (Thrombus)
For Use With (Application) Immunohistochemistry-Paraffin
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Thrombus
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.