missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph Instruments Heat-On Inserts
Suitable for heating round bottom flasks
128.00 € - 218.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11731659
|
Heidolph Instruments
505-80061-00 |
10 mL |
132.00 €
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
10319943
|
Heidolph Instruments
505-80062-00 |
25 mL |
129.00 €
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
10432915
|
Heidolph Instruments
505-80065-00 |
150 mL |
218.00 €
Each |
Estimated Shipment: 24-12-2024
Log in to see stock. |
|||||
10617974
|
Heidolph Instruments
505-80063-00 |
50 mL |
128.00 €
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
Description
- For optimal use it is recommended that you also purchase a Multi-Well holder and safety lifting handles
- The maximum temperature for continuous use is 260°C
- To measure this precisely with a temperature sensor, a small hole is provided on one side of the Heat-On Block
Specifications
100/230 V | |
Fluoropolymer | |
Aluminium 6082 HE30 |