missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph Instruments Heat-On Multi-Well Holder
This unique Heat-On Multi-Well Holder is designed to hold either one or two inserts for flasks that also features cut-away sides for use with two or three neck flasks and accept the optional safety lifting handles.
Brand: Heidolph Instruments 505-80060-00
This item is not returnable.
View return policy
Description
- This unique Multi-Well Holder is designed to hold either one or two inserts for flasks (only accepts one 150 mL insert)
- The inserts are available for 10 mL, 25 mL, 50 mL, 100 mL and 150 mL flasks
- Flask inserts also feature cut-away sides for use with two or three neck flasks and accept the optional safety lifting handles

Specifications
Multi-Well Holder | |
260°C | |
Fluoropolymer |
500°F | |
Block Adapter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heidolph Instruments Heat-On Multi-Well Holder > Multi-Well Holder; Temperature: 260°C (500°F)
Spot an opportunity for improvement?Share a Content Correction