missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1073B Tyvek Autoclave Pouch, with Steam Indicator Dot
BIOPHARMAGREENER_CHOICE

Tyvek Pouch that offers high breathability while providing a barrier from microbial contamination after sterilization.

Brand:  Keystone Cleanroom Products COVMGTVKUB1X14

 View more versions of this product

Product Code. 17718357

  • 417.00 € / Pack of 100
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • All Tyvek construction for strength and microbial barrier
  • Designed for steam sterilization
  • Strong Factory seals
  • Heat Seal Closure
  • Recommended for sterilization of pharmaceutical equipment and Components
  • Manufactured and packaged in a ISO 6 cleanroom
  • Improves the efficiency of the Parts Preparation Department
Specifications

Specifications

Autoclavable
200 mL
Drawstring
Single-use
Pharmaceutical Equipment
Non-sterile
<250°F
35.6 x 2.54 cm (14 x 1 in.)
35.6 cm
2.54 cm
7 oz.
COC
White
Steam Sterilization
Tyvek
Steam
<125°C
14 in.
1 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.