Learn More
Keystone Cleanroom Products 1073B Tyvek Autoclave Pouch, with Steam Indicator Dot
GREENER_CHOICE
Tyvek Pouch that offers high breathability while providing a barrier from microbial contamination after sterilization.
Brand:  Keystone Cleanroom Products COVMGTVKUB3X6
 View more versions of this product
Product Code. 17718397
  • 361.00 € / Pack of 100
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All Tyvek construction for strength and microbial barrier
  • Designed for steam sterilization
  • Strong Factory seals
  • Heat Seal Closure
  • Recommended for sterilization of pharmaceutical equipment and Components
  • Manufactured and packaged in a ISO 6 cleanroom
  • Improves the efficiency of the Parts Preparation Department
Specifications
Specifications
Autoclavable
COC
White
Steam Sterilization
Tyvek
Steam
<125°C
6 in.
3 in.
12 oz.
Heat Seal
Single-use
Pharmaceutical Equipment
Non-sterile
<250°F
15.2 x 7.6 cm (6 x 3 in.)
15.2 cm
7.6 cm