missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1073B Tyvek Autoclave Pouch, with Steam Indicator Dot
BIOPHARMAGREENER_CHOICE

Tyvek Pouch that offers high breathability while providing a barrier from microbial contamination after sterilization.

Brand:  Keystone Cleanroom Products COVMGTVKUB36X60

 View more versions of this product

Product Code. 17768387

  • 1295.00 € / Pack of 50
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • All Tyvek construction for strength and microbial barrier
  • Designed for steam sterilization
  • Strong Factory seals
  • Heat Seal Closure
  • Recommended for sterilization of pharmaceutical equipment and Components
  • Manufactured and packaged in a ISO 6 cleanroom
  • Improves the efficiency of the Parts Preparation Department
Specifications

Specifications

Autoclavable
62.1 L
Heat Seal
Single-use
Pharmaceutical Equipment
Non-sterile
<250°F
152.4 x 91.4 cm (60 x 36 in.)
152.4 cm
91.4 cm
1200 oz.
COC
White
Steam Sterilization
Tyvek
Steam
<125°C
60 in.
36 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers