Learn More
Keystone Cleanroom Products 1073B Tyvek Autoclave Pouch, with Steam Indicator Dot
BIOPHARMAGREENER_CHOICE
Tyvek Pouch that offers high breathability while providing a barrier from microbial contamination after sterilization.
Brand:  Keystone Cleanroom Products COVMGTVKUB24X36
 View more versions of this product
Product Code. 17788357
  • 713.00 € / Pack of 50
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All Tyvek construction for strength and microbial barrier
  • Designed for steam sterilization
  • Strong Factory seals
  • Heat Seal Closure
  • Recommended for sterilization of pharmaceutical equipment and Components
  • Manufactured and packaged in a ISO 6 cleanroom
  • Improves the efficiency of the Parts Preparation Department
Specifications
Specifications
White
Pharmaceutical Equipment
Tyvek
61 x 91.4 cm (24 x 36 in.)