Learn More
Keystone Cleanroom Products Heat Seal Tyvek™ Steam Sterilization Pouches
BIOPHARMAGREENER_CHOICE
Heat Seal Tyvek™ Steam Sterilization Pouches provide a strong barrier from microbial contamination.
Brand:  Keystone Cleanroom Products BBMGTVKHS2424
 View more versions of this product
Product Code. 17787817
  • 4145.00 € / Pack of 250
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing a strong barrier from microbial contamination
  • The clear film side offers high clarity along with strength and durability
  • Should be heat sealed closed using a validate-able impulse, or continuous band heat sealer
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Works with both Impulse and Continuous Band Sealers
Specifications
Specifications
Autoclave Bag
Certificate of Conformance (COC)
White
Steam Sterilization
Non-sterile
250/Pk.
24 in.
24 in.
Autoclavable
Heat Seal
Single-use
Tyvek
Yes
24 x 24 in.
61 cm
61 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Keystone Cleanroom Products Heat Seal Tyvek™ Steam Sterilization Pouches > Steam Sterilization Pouches; Dimensions (L x W): 24 x 24 in.