Learn More
Eppendorf™ Heat Sealer S200
Provides user-friendly and reliable hermetic sealing of a variety of multiwell plates.
Brand:  Eppendorf™ 5392000030
 View more versions of this product
Product Code. 15785665
  • 4492.90 € / Each
Explore more special offers
Description
Description
  • Eliminates evaporation in PCR
  • Adjustable sealing temperatures for more flexibility
  • Seals 96- and 384-well plates
  • Sealing time selectable from 1 to 9 seconds in 0.5 second increments
  • Integrated thermostat prevents overheating
  • Powerful handle for ergonomic operation
  • PCR plate adapter included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/60 Hz
220 × 321 × 425 mm
96- and 384-well plates
125°C to 200°C in 1°C increments
100/230 V
100/230 V, 50/60 Hz
GB Plug