Learn More
IKA™ Heating Block
IKA Flow Heating Block holds four disc reactors which are fed by the IKA FLOW during reaction.
Brand:  IKA™ 0020103369
Product Code. 17593636
  • 305.00 € / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • IKA Flow Heating Block is installed to the magnetic stirrer RCT digital and has the maximum capacity of holding four disc reactors which are fed by the IKA FLOW during reaction.
  • Disc reactos can be purchased separately.
  • Scope of delivery
    • Heating Block
Specifications
Specifications
Magnetic Stirrer RCT Digital