Learn More
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations Product Code.: 11894770
Estimated Shipment: 05-08-2025
to see stock.
Add to basket
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations
Product Code. 11894770 Shop All Cole-Parmer Products
4 x 96-well PTFE Microplates
Click to view available options
Holds:
4 x 96-well PTFE Microplates
40 x 24 mm Tubes
96 x 16 mm Tubes
Product Code. 11894770
Brand: Cole-Parmer™ 9996805
Estimated Shipment: 05-08-2025
to see stock.
Add to basket
Add to basket
Accessory heating blocks to keep your reactions running smoothly.
  • Compatible with Electrothermal RS9000 Reaction Stations
One year warranty
  • Specifications
    Description Heating Block for Reaction System
    For Use With (Equipment) RS-250D-S Shaking Reaction Stations
    Holds 4 x 96-well PTFE Microplates
    Material PTFE
    Type Heating Block
    Includes 4 Blocks, 96 Wells
    Warranty 1 Year
    Product Content Correction
    Product Title
    Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations >