Learn More
Grant Instruments™ Heating Block for TS-DW Deep-well Microplate Thermoshaker
Click to view available options
For Use With (Application):
Axygen™ 96/2200µl deep-well plate
Eppendorf™ 96/1000µl deep-well plate
Sarstedt™ 96/2200µl deep-well plate
Starlab™ 96/1200µl deep-well plate
Description
Block for one deep-well plate
- Shaped to fit the microplates for better heat transfer
- Three year warranty
Specifications
Specifications
For Use With (Equipment) | TS-DW Microplate Thermoshaker |
Type | Block |
For Use With (Application) | Eppendorf™ 96/1000µl deep-well plate |