Learn More
Grant Instruments™ Heating Block for TS-DW Deep-well Microplate Thermoshaker
Product Code. 15450474
Click to view available options
For Use With (Application):
Axygen™ 96/2200µl deep-well plate
Eppendorf™ 96/1000µl deep-well plate
Sarstedt™ 96/2200µl deep-well plate
Starlab™ 96/1200µl deep-well plate
Accommodates one deep-well plate for TS-DW Deep-well Microplate Thermoshaker
Block for one deep-well plate
  • Shaped to fit the microplates for better heat transfer
  • Three year warranty
Specifications
For Use With (Equipment) TS-DW Microplate Thermoshaker
Type Block
For Use With (Application) Axygen™ 96/2200µl deep-well plate