Learn More
Electrothermal™ Heating Controller Temperature Probes
Choose the appropriate temperature probe specific to the model of Electrothermal Digital Heating Controller (sold separately)
Brand:  Electrothermal™ 99965-03
Product Code. 11544098
  • 740.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Temperature Probes
  • Suitable for use with Electrothermal Digital Heating Controllers
  • Controller sold separately
Specifications
Specifications
752°F
PT100 Probe
Temperature Probe
400°C
Class A
MC810B Digital Controller