missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heating/Cooling Mini Shaker for PCR

Ideal for analyses in a stable, controlled environment and PCR plate ready without the need for any additional accessories

Brand:  Fisherbrand™ 88861040

2293.27 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 15574070

  • 2955.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: PCR block, Polycarbonate lid
This item is not returnable. View return policy

Description

Description

 

  • Microprocessor control - The variable speed microprocessor control provides consistent, uniform shaking action. Microprocessor will display last set-point and will restart if power is interrupted.
  • PID temperature controller - Maintains precise temperature control from 10°C below ambient to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C increments.
  • Triple eccentric drive - Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation.
  • LED display - Touch pad controls with easy-to-read, independent LED displays for temperature, speed and time allow operator to view all settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
  • Temperature calibration mode - Allows user to calibrate unit to an external temperature device
  • Applications - ELISA, Enzyme Activity Analysis, Enzyme cutting, DNA ligation, PCR

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 mm
230 V,1 A, 450 W, Plug Type: EU, UK, CN
Ambient -10° to 95°C (Lowest Possible Temp: 5°C)
EU, UK, CN Plug
278 x 222 x 155 mm
415 x 293 x 308 mm
LED
1 sec. to 9999 min.
150 to 1200 rpm (PCR plate)
230 V
CE
Aluminum Platform
7 x 5.5 in. (180 x 140 mm)
Audible and Visual
PCR block, Polycarbonate lid
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.