missing translation for 'onlineSavingsMsg'
Learn More

Electrothermal™ RE0100 100mL Replacement Cartridge for Heating Mantle

Helps to ensure that the temperature in a heating mantle is uniformly distributed across the bottom.

Brand:  Electrothermal™ 03012-25

Product Code. 11718412

  • 313.00 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For 100mL flask capacity
  • Supplied individually
  • Suitable for EM, EM/E, EM/C, EM/CE series only
Specifications

Specifications

100mL flask capacity
230 V
Heating Mantle Element
EM Series Heating Mantles
60w
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers