missing translation for 'onlineSavingsMsg'
Learn More

Gestigkeit Harry Heating Unit

Each SG hotplate is equipped with an output controller allowing variable adjustment of the heating output from 10 to 100%.

Brand:  Gestigkeit Harry SG6

 View more versions of this product

Product Code. 10639562

  • 2080.00 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Series and supporting rod hot plates
  • Smoothly finished, cast-iron, hot plate with concave central expansion recess for warp free operation. All housing parts are constructed from stainless steel and each unit has a screw-on fitting for a supporting rod.
  • The appliances have been specifically designed for non-stop use.
  • Regulation of model
    • Each unit has a heating output controller that enables a variable adjustment between 10 to 100%.
Specifications

Specifications

Stainless Steel
Hotplate
230 V
500°C
3000 W
1055 x 210 x 160 mm
6
Cast Steel
14.8 kg
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.