missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEATR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561371
This item is not returnable.
View return policy
Description
Immunogen sequence: QELFHQFVSL STSGGKYQFL ADSDTSLMLS LNHPLAPVRI LAMNHLKKIM KTSKEGVDES FIKEAVLARL GDDNIDVVLS AISAFEIFKE HFS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.
Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt]Specifications
HEATR1 | |
Polyclonal | |
Unconjugated | |
HEATR1 | |
AA517551; B130016L12Rik; BAP28; BC019693; FLJ10359; HEAT repeat containing 1; HEAT repeat-containing protein 1; HEAT repeat-containing protein 1, N-terminally processed; HEATR1; Protein BAP28; protein UTP10; RGD1564769; U3 small nucleolar RNA-associated protein 10 homolog; UTP10; UTP10, small subunit (SSU) processome component, homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H583 | |
HEATR1 | |
Recombinant protein corresponding to Human HEATR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only