Learn More
Cole-Parmer™ Heavy-Duty Aluminum Foil
This Heavy-Duty Aluminum Foil is an all-purpose foil used for freezer storage, wrapping, sealing, autoclaving, lining, and weighing or transferring samples.
Brand:  Cole-Parmer™ 06275-05
Product Code. 11720109
  • 360.88 € / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Packaged in a dispenser box with serrated cutting strip.
Specifications
Specifications
Freezer storage, wrapping, sealing, autoclaving, lining, and weighing or transferring samples
Heavy-Duty Aluminum Foil
45.7 cm
Aluminum
152.4 m
18 in.
500 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Heavy-Duty Aluminum Foil > Heavy-Duty Aluminum Foil; Length: 152.4 m (500 ft.); Width: 45.7 cm (18 in.)