Learn More
3M™ Heavy Duty EPDM Breathing Tube
Extended fixed-length breathing tube made from EPDM rubber.
51.70 € - 60.60 €
Specifications
Material EPDM Rubber
Product Type Breathing Tube
View More Specs
Products 2
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
17786993
View Documents
3M™
7100265243
SC-BT-44-L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17796993
View Documents
3M™
7100265244
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Suitable for use with specific 3M™ Versaflo™ headtops including M-200, M-300 and S Series Hoods.
  • EPDM rubber hose
  • 100 cm length
  • Protective breathing tube covers available to lengthen service life of breathing tubes
Specifications
Specifications
EPDM Rubber
Breathing Tube