Learn More
Liebherr™ Heavy-Duty Laboratory Refrigerator
Brand:  Liebherr™ LKPv 1420
Product Code. 10094084
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
A range of purpose built laboratory refrigerators, single and double door, solid and glass door.
  • Data logging of temperature every two minutes
  • Audible and visual door-open and hi-low temperature alarm
  • Volt free contact for connection to remote system
  • Data transferable to external unit
Capacity: 1427LTemperature Range: -2 to 16°CMaterial: Stainless SteelDisplay Type: External digitalTotal Shelves: 8Surface Coating: PlasticShelf Count: 8Door Count: 2Defrost Type: AutomaticDoor Type: Self closing doorIncludes: Alarms, data logger and castors, door lock, electronic controllerInterior Height: 1550mmInterior Width: 125mmExterior Dimensions: 2150 H x 1430 W x 830 DmmWeight: 199kgShipping Weight: 254kgExterior Width: 1430mmInterior Depth: 700mmExterior Height: 2150mmDimensions (D x W x H) Interior: 700 x 1250 x 1550mmDimensions (L x W x H) Exterior: 1430 W x 2150 HmmVoltage: 220 to 240VHertz: 50HzElectrical Requirements: 220 to 240V, 50Hz